siRNA Details
| Gene Symbol: LOC107984282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017017028.1 | XP_016872517.1 | 3 | 7176 | ||
| GenBank mRNA | AF086154.1 | - | 2 | 569 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107984282| Gene Symbol: LOC107984282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017017028.1 | XP_016872517.1 | 3 | 7176 | ||
| GenBank mRNA | AF086154.1 | - | 2 | 569 | ||