siRNA Details
| Gene Symbol: GUSBP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024448.2 | 11, 12 | 2967 | |||
| GenBank mRNA | AK056875.1 | - | 11, 12 | 2897 | ||
| AK125808.1 | - | 4, 5 | 3367 | |||
| BC063045.1 | - | 9, 10 | 1210 | |||
| X52204.1 | - | 3, 4 | 461 | |||

GUSBP11
Genomic Map