siRNA Details
| Gene Symbol: GUSBP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024448.2 | 7 | 1100 | |||
| GenBank mRNA | AK056875.1 | - | 7 | 1033 | ||
| BC063045.1 | - | 7 | 863 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUSBP11| Gene Symbol: GUSBP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024448.2 | 7 | 1100 | |||
| GenBank mRNA | AK056875.1 | - | 7 | 1033 | ||
| BC063045.1 | - | 7 | 863 | |||