siRNA Details
| Gene Symbol: LOC101929524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109946.1 | 4 | 1601 | |||
| GenBank mRNA | AK097784.1 | - | 4 | 1599 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929524| Gene Symbol: LOC101929524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109946.1 | 4 | 1601 | |||
| GenBank mRNA | AK097784.1 | - | 4 | 1599 | ||