siRNA Details
| Gene Symbol: LINC01532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040036.1 | 5 | 1841 | |||
| GenBank mRNA | AK055100.1 | - | 5 | 1839 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01532| Gene Symbol: LINC01532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040036.1 | 5 | 1841 | |||
| GenBank mRNA | AK055100.1 | - | 5 | 1839 | ||