siRNA Details
| Gene Symbol: ERVH48-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC005107.1 | - | 2 | 2397 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF090920.1 | - | 1 | 484 | Chr. 11 - 7514778 - 7517446 [-] on Build GRCh38 | |

ERVH48-1
Genomic Map