siRNA Details
Gene Symbol: LINC01262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121679.1 | 2 | 219 | |||
GenBank mRNA | BC092460.1 | - | 2 | 219 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD641470.1 | - | 222 | Chr. - - [-] on Build |
