siRNA Details
| Gene Symbol: ESRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027122.1 | 4 | 2477 | |||
| GenBank mRNA | DQ445779.2 | - | 4 | 2476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ESRG| Gene Symbol: ESRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027122.1 | 4 | 2477 | |||
| GenBank mRNA | DQ445779.2 | - | 4 | 2476 | ||