siRNA Details
| Gene Symbol: UBAC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036531.1 | 2 | 917 | |||
| NR_036532.1 | 2 | 643 | ||||
| GenBank mRNA | AF339765.1 | - | 1 | 115 | ||
| AK097488.1 | - | 2 | 922 | |||
| BC043515.1 | - | 2 | 598 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI827524.1 | - | 2 | 598 | Chr. 13 - 99196953 - 99200710 [-] on Build GRCh38 | |

UBAC2-AS1
Genomic Map