siRNA Details
| Gene Symbol: UBAC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036531.1 | 2 | 1332 | |||
| NR_036532.1 | 2 | 1058 | ||||
| GenBank mRNA | AF339765.1 | - | 1 | 530 | ||
| AK097488.1 | - | 2 | 1337 | |||
| BC043515.1 | - | 2 | 1013 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBAC2-AS1| Gene Symbol: UBAC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036531.1 | 2 | 1332 | |||
| NR_036532.1 | 2 | 1058 | ||||
| GenBank mRNA | AF339765.1 | - | 1 | 530 | ||
| AK097488.1 | - | 2 | 1337 | |||
| BC043515.1 | - | 2 | 1013 | |||