siRNA Details
| Gene Symbol: LOC100507657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134581.1 | 1 | 3528 | |||
| GenBank mRNA | CR627208.1 | - | 1 | 3527 | ||
| CR936633.1 | - | 1 | 3527 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507657| Gene Symbol: LOC100507657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134581.1 | 1 | 3528 | |||
| GenBank mRNA | CR627208.1 | - | 1 | 3527 | ||
| CR936633.1 | - | 1 | 3527 | |||