siRNA Details
| Gene Symbol: UPK1A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046420.1 | 2 | 463 | |||
| GenBank mRNA | BC007817.1 | - | 2 | 472 | ||
| BF683645.1 | - | 2 | 463 | |||
| BX283291.1 | - | 2 | 464 | |||
| EL733629.1 | - | 2 | 423 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UPK1A-AS1| Gene Symbol: UPK1A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046420.1 | 2 | 463 | |||
| GenBank mRNA | BC007817.1 | - | 2 | 472 | ||
| BF683645.1 | - | 2 | 463 | |||
| BX283291.1 | - | 2 | 464 | |||
| EL733629.1 | - | 2 | 423 | |||