siRNA Details
| Gene Symbol: LINC01139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015407.1 | 2 | 967 | |||
| GenBank mRNA | BC044843.1 | - | 2 | 967 | ||
| BC064529.1 | - | 2 | 973 | |||
| BC073894.1 | - | 2 | 944 | |||
| BC113688.1 | - | 1 | 683 | |||
| BC113690.1 | - | 1 | 683 | |||

LINC01139
Genomic Map