siRNA Details
| Gene Symbol: LOC284933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038917.1 | 3 | 568 | |||
| GenBank mRNA | BC036359.1 | - | 3 | 568 | ||
| BC048428.1 | - | 2 | 588 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI830326.1 | - | 3 | 568 | Chr. 22 - 48544684 - 48547387 [-] on Build GRCh38 | |

LOC284933
Genomic Map