siRNA Details
| Gene Symbol: LOC105378663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135048.1 | 2 | 237 | |||
| GenBank mRNA | BX115242.1 | - | 2 | 188 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY050812.1 | - | 2 | 233 | Chr. 1 - 38873405 - 38876197 [+] on Build GRCh38 | |

LOC105378663
Genomic Map