siRNA Details
| Gene Symbol: LOC101928063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110727.1 | 3 | 1864 | |||
| GenBank mRNA | AK090553.1 | - | 2 | 2192 | ||
| AK094825.1 | - | 3 | 1827 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928063| Gene Symbol: LOC101928063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110727.1 | 3 | 1864 | |||
| GenBank mRNA | AK090553.1 | - | 2 | 2192 | ||
| AK094825.1 | - | 3 | 1827 | |||