siRNA Details
| Gene Symbol: LINC01527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042056.1 | - | 2 | 470 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM545658.1 | - | 585 | Chr. - - [-] on Build | ||

LINC01527
Genomic Map