siRNA Details
| Gene Symbol: ANKRD44-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046837.1 | 5 | 862 | |||
| GenBank mRNA | BC042827.1 | - | 5 | 862 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD44-IT1| Gene Symbol: ANKRD44-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046837.1 | 5 | 862 | |||
| GenBank mRNA | BC042827.1 | - | 5 | 862 | ||