siRNA Details
| Gene Symbol: LINC00837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038374.1 | 3 | 3914 | |||
| NR_038375.1 | 2 | 3812 | ||||
| NR_038376.1 | 2 | 3691 | ||||
| GenBank mRNA | AL080132.1 | - | 1 | 2634 | ||
| BC028422.1 | - | 2 | 3735 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB524548.1 | - | 1 | 381 | Chr. 10 - 28789313 - 28789791 [-] on Build GRCh38 | |

LINC00837
Genomic Map