siRNA Details
| Gene Symbol: LINC00837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038374.1 | 3 | 443 | |||
| NR_038375.1 | 2 | 341 | ||||
| NR_038376.1 | 2 | 220 | ||||
| GenBank mRNA | BC028422.1 | - | 2 | 268 | ||
| DB072413.1 | - | 2 | 221 | |||
| DB091920.1 | - | 3 | 366 | |||
| DB099527.1 | - | 2 | 177 | |||

LINC00837
Genomic Map