siRNA Details
| Gene Symbol: LOC284600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056486.1 | - | 2 | 1015 | ||
| BC000227.1 | - | 1 | 477 | |||
| BC006442.1 | - | 1 | 477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284600| Gene Symbol: LOC284600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056486.1 | - | 2 | 1015 | ||
| BC000227.1 | - | 1 | 477 | |||
| BC006442.1 | - | 1 | 477 | |||