siRNA Details
| Gene Symbol: DPP9-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242901.1 | NP_001229830.1 | 3 | 1779 | ||
| GenBank mRNA | AK126095.1 | - | 3 | 1779 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPP9-AS1| Gene Symbol: DPP9-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242901.1 | NP_001229830.1 | 3 | 1779 | ||
| GenBank mRNA | AK126095.1 | - | 3 | 1779 | ||