siRNA Details
| Gene Symbol: CHODL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024354.1 | 1 | 133 | |||
| GenBank mRNA | T74237.1 | - | 1 | 133 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHODL-AS1| Gene Symbol: CHODL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024354.1 | 1 | 133 | |||
| GenBank mRNA | T74237.1 | - | 1 | 133 | ||