siRNA Details
| Gene Symbol: CCDC13-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098828.1 | - | 3 | 1067 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC13-AS1| Gene Symbol: CCDC13-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098828.1 | - | 3 | 1067 | ||