siRNA Details
| Gene Symbol: LOC100506937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134332.1 | 2 | 654 | |||
| GenBank mRNA | BX103078.1 | - | 2 | 654 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506937| Gene Symbol: LOC100506937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134332.1 | 2 | 654 | |||
| GenBank mRNA | BX103078.1 | - | 2 | 654 | ||