siRNA Details
| Gene Symbol: LOC105375483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133947.1 | 5 | 1289 | |||
| GenBank mRNA | BC041947.1 | - | 5 | 1289 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105375483| Gene Symbol: LOC105375483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133947.1 | 5 | 1289 | |||
| GenBank mRNA | BC041947.1 | - | 5 | 1289 | ||