siRNA Details
| Gene Symbol: LOC100129434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125368.1 | 3 | 534 | |||
| GenBank mRNA | AK096196.1 | - | 3 | 746 | ||
| AK295617.1 | - | 4 | 1311 | |||
| AK311113.1 | - | 3 | 534 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA104205.1 | - | 3 | 535 | Chr. 2 - 56175832 - 56185770 [-] on Build GRCh38 | |

LOC100129434
Genomic Map