siRNA Details
| Gene Symbol: LOC101928008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120539.1 | 3 | 454 | |||
| GenBank mRNA | U80769.1 | - | 3 | 466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928008| Gene Symbol: LOC101928008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120539.1 | 3 | 454 | |||
| GenBank mRNA | U80769.1 | - | 3 | 466 | ||