siRNA Details
Gene Symbol: FENDRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033925.1 | 4 | 2954 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FENDRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033925.1 | 4 | 2954 |