siRNA Details
| Gene Symbol: EWSAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026949.1 | 3 | 1226 | |||
| GenBank mRNA | AK091087.1 | - | 3 | 1214 | ||
| AK097560.1 | - | 4 | 1055 | |||
| AK292174.1 | - | 3 | 779 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EWSAT1| Gene Symbol: EWSAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026949.1 | 3 | 1226 | |||
| GenBank mRNA | AK091087.1 | - | 3 | 1214 | ||
| AK097560.1 | - | 4 | 1055 | |||
| AK292174.1 | - | 3 | 779 | |||