siRNA Details
Gene Symbol: LOC101928211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110180.1 | 4 | 516 | |||
GenBank mRNA | BC031318.2 | - | 4 | 520 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI458366.1 | - | 4 | 525 | Chr. 7 - 124337380 - 124352098 [-] on Build GRCh38 |
