siRNA Details
Gene Symbol: LINC01550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015430.1 | 1 | 139 | |||
GenBank mRNA | AK091668.1 | - | 1 | 125 | ||
BC043585.1 | - | 1 | 139 | |||
BX647564.1 | - | 1 | 123 | |||
JX073283.1 | - | 1 | 75 |
