siRNA Details
Gene Symbol: LINC01550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015430.1 | 2 | 400 | |||
GenBank mRNA | AK091668.1 | - | 2 | 386 | ||
AK097943.1 | - | 2 | 255 | |||
BC043585.1 | - | 2 | 400 | |||
BX647564.1 | - | 2 | 384 | |||
JX073283.1 | - | 2 | 336 |
