siRNA Details
Gene Symbol: LOC105369860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135021.1 | 2 | 1079 | |||
GenBank mRNA | BC034620.1 | - | 2 | 1051 | ||
BC045810.1 | - | 2 | 1079 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105369860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135021.1 | 2 | 1079 | |||
GenBank mRNA | BC034620.1 | - | 2 | 1051 | ||
BC045810.1 | - | 2 | 1079 |