siRNA Details
| Gene Symbol: LINC00707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038291.1 | 5 | 1312 | |||
| GenBank mRNA | AK094154.1 | - | 5 | 1318 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00707| Gene Symbol: LINC00707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038291.1 | 5 | 1312 | |||
| GenBank mRNA | AK094154.1 | - | 5 | 1318 | ||