siRNA Details
| Gene Symbol: LINC00911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102737.1 | 3 | 514 | |||
| NR_102738.1 | 2 | 404 | ||||
| GenBank mRNA | AK131026.1 | - | 3 | 514 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00911| Gene Symbol: LINC00911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102737.1 | 3 | 514 | |||
| NR_102738.1 | 2 | 404 | ||||
| GenBank mRNA | AK131026.1 | - | 3 | 514 | ||