siRNA Details
| Gene Symbol: ADORA2A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028483.2 | 4 | 890 | |||
| GenBank mRNA | BC045098.1 | - | 4 | 631 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADORA2A-AS1| Gene Symbol: ADORA2A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028483.2 | 4 | 890 | |||
| GenBank mRNA | BC045098.1 | - | 4 | 631 | ||