siRNA Details
| Gene Symbol: KCNK4-TEX40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133662.1 | 7 | 1638 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNK4-TEX40| Gene Symbol: KCNK4-TEX40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133662.1 | 7 | 1638 | |||