siRNA Details
| Gene Symbol: UST-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038408.1 | 3 | 526 | |||
| GenBank mRNA | BC034817.1 | - | 3 | 526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UST-AS1| Gene Symbol: UST-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038408.1 | 3 | 526 | |||
| GenBank mRNA | BC034817.1 | - | 3 | 526 | ||