siRNA Details
Gene Symbol: LOC101929488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125818.1 | 9 | 1675 | |||
GenBank mRNA | BC039330.1 | - | 9 | 1675 | ||
BC048118.1 | - | 9 | 1674 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125818.1 | 9 | 1675 | |||
GenBank mRNA | BC039330.1 | - | 9 | 1675 | ||
BC048118.1 | - | 9 | 1674 |