siRNA Details
| Gene Symbol: LOC101929488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125818.1 | 3 | 596 | |||
| GenBank mRNA | BC039330.1 | - | 3 | 596 | ||
| BC048118.1 | - | 3 | 595 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929488| Gene Symbol: LOC101929488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125818.1 | 3 | 596 | |||
| GenBank mRNA | BC039330.1 | - | 3 | 596 | ||
| BC048118.1 | - | 3 | 595 | |||