siRNA Details
Gene Symbol: RASSF8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038227.1 | 3, 4 | 371 | |||
NR_038228.1 | 3, 4 | 358 | ||||
GenBank mRNA | AK090817.1 | - | 3, 4 | 272 | ||
BC004962.1 | - | 3, 4 | 371 | |||
DA306887.1 | - | 3, 4 | 282 |
