siRNA Details
| Gene Symbol: PTOV1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040037.1 | 4 | 1272 | |||
| GenBank mRNA | AK092213.1 | - | 3 | 2886 | ||
| BC043359.1 | - | 4 | 1272 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTOV1-AS1| Gene Symbol: PTOV1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040037.1 | 4 | 1272 | |||
| GenBank mRNA | AK092213.1 | - | 3 | 2886 | ||
| BC043359.1 | - | 4 | 1272 | |||