siRNA Details
| Gene Symbol: TUG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002323.2 | 2 | 2845 | |||
| NR_110492.1 | 3 | 2901 | ||||
| NR_110493.1 | 2 | 2845 | ||||
| GenBank mRNA | AK000625.1 | - | 2 | 844 | ||
| AK000749.1 | - | 3 | 745 | |||
| AK056480.1 | - | 2 | 1379 | |||
| AK057695.1 | - | 1 | 869 | |||
| BC037986.1 | - | 2 | 1002 | |||

TUG1
Genomic Map