siRNA Details
| Gene Symbol: TUG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002323.2 | 2 | 2804 | |||
| NR_110492.1 | 3 | 2860 | ||||
| NR_110493.1 | 2 | 2804 | ||||
| GenBank mRNA | AK000625.1 | - | 2 | 803 | ||
| AK000749.1 | - | 3 | 704 | |||
| AK056480.1 | - | 2 | 1338 | |||
| AK057695.1 | - | 1 | 828 | |||
| BC037986.1 | - | 2 | 961 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA834152.1 | - | 1 | 38 | Chr. 22 - 30973052 - 30975455 [+] on Build GRCh38 | |

TUG1
Genomic Map