siRNA Details
| Gene Symbol: MIAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003491.3 | 5 | 10046 | |||
| NR_033319.2 | 4 | 9920 | ||||
| NR_033320.2 | 5 | 9972 | ||||
| NR_033321.2 | 4 | 9846 | ||||
| GenBank mRNA | AB263414.1 | - | 5 | 10007 | ||
| AB263415.1 | - | 4 | 9881 | |||
| AB263416.1 | - | 5 | 9933 | |||
| AB263417.1 | - | 4 | 9807 | |||
| AK098833.1 | - | 4 | 1578 | |||
| AK127256.1 | - | 1 | 5158 | |||

MIAT
Genomic Map