siRNA Details
| Gene Symbol: LOC105371335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135171.1 | 3 | 364 | |||
| NR_135172.1 | 2 | 242 | ||||
| GenBank mRNA | HY013069.1 | - | 2 | 243 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105371335| Gene Symbol: LOC105371335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135171.1 | 3 | 364 | |||
| NR_135172.1 | 2 | 242 | ||||
| GenBank mRNA | HY013069.1 | - | 2 | 243 | ||