siRNA Details
| Gene Symbol: SUCLG2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109992.1 | 1 | 365 | |||
| NR_109993.1 | 1 | 365 | ||||
| GenBank mRNA | AL832184.1 | - | 1 | 305 | ||
| BC038210.1 | - | 1 | 365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SUCLG2-AS1| Gene Symbol: SUCLG2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109992.1 | 1 | 365 | |||
| NR_109993.1 | 1 | 365 | ||||
| GenBank mRNA | AL832184.1 | - | 1 | 305 | ||
| BC038210.1 | - | 1 | 365 | |||