siRNA Details
| Gene Symbol: LOC101928844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110740.1 | 1 | 293 | |||
| GenBank mRNA | BC033124.1 | - | 1 | 293 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE299888.1 | - | 1 | 269 | Chr. 19 - 5847489 - 5857902 [+] on Build GRCh38 | |

LOC101928844
Genomic Map