siRNA Details
| Gene Symbol: LOC101928449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110089.1 | 1 | 205 | |||
| GenBank mRNA | BC031963.1 | - | 1 | 205 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928449| Gene Symbol: LOC101928449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110089.1 | 1 | 205 | |||
| GenBank mRNA | BC031963.1 | - | 1 | 205 | ||